Assay Details
| Gene Symbol: rgs-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307891.3 | NP_001294820.1 | 5 - 6 | 645 | 88 | |
| NM_001307892.4 | NP_001294821.1 | 3 - 4 | 802 | 88 | ||
More Information
Set Membership: |
|

rgs-6
Genomic Map