Assay Details
| Gene Symbol: tag-294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076566.4 | NP_508967.1 | 2 - 3 | 301 | 127 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tag-294| Gene Symbol: tag-294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076566.4 | NP_508967.1 | 2 - 3 | 301 | 127 | |
Set Membership: |
|