Assay Details
| Gene Symbol: puf-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076579.8 | NP_508980.2 | 6 - 7 | 1312 | 71 | |
| Gene Symbol: atat-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383590.1 | 5 - 6 | 1214 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383590.1 transcripts.
Set Membership: |
|

puf-9
Genomic Map