Assay Details
Gene Symbol: kin-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083302.4 | NP_001076771.1 | 2 - 3 | 206 | 108 | |
NM_001380985.1 | NP_001367669.1 | 2 - 3 | 190 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
