Assay Details
| Gene Symbol: T10E10.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001375049.2 | NP_001361925.1 | 9 | 1181 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

T10E10.4| Gene Symbol: T10E10.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001375049.2 | NP_001361925.1 | 9 | 1181 | 58 | |
Set Membership: |
|