Assay Details
Gene Symbol: smf-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076730.3 | NP_509131.2 | 10 - 11 | 1534 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
