Assay Details
| Gene Symbol: smf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029621.5 | NP_001024792.2 | 8 - 9 | 1298 | 93 | |
| NR_132544.1 | 7 - 8 | 1045 | 93 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

smf-1| Gene Symbol: smf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029621.5 | NP_001024792.2 | 8 - 9 | 1298 | 93 | |
| NR_132544.1 | 7 - 8 | 1045 | 93 | |||
Set Membership: |
|