Assay Details
Gene Symbol: smf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029621.5 | NP_001024792.2 | 9 - 10 | 1405 | 84 | |
NR_132544.1 | 8 - 9 | 1152 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
