Assay Details
| Gene Symbol: dhs-28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076745.7 | NP_509146.1 | 3 - 4 | 623 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dhs-28| Gene Symbol: dhs-28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076745.7 | NP_509146.1 | 3 - 4 | 623 | 70 | |
Set Membership: |
|