Assay Details
| Gene Symbol: rig-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076754.5 | NP_509155.1 | 7 - 8 | 1276 | 141 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rig-3| Gene Symbol: rig-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076754.5 | NP_509155.1 | 7 - 8 | 1276 | 141 | |
Set Membership: |
|