Assay Details
| Gene Symbol: ubc-17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076787.5 | NP_509188.3 | 12 - 13 | 1931 | 124 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ubc-17| Gene Symbol: ubc-17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076787.5 | NP_509188.3 | 12 - 13 | 1931 | 124 | |
Set Membership: |
|