Assay Details
| Gene Symbol: ZK867.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001368523.1 | NP_001355384.1 | 4 - 5 | 490 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZK867.2| Gene Symbol: ZK867.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001368523.1 | NP_001355384.1 | 4 - 5 | 490 | 97 | |
Set Membership: |
|