Assay Details
| Gene Symbol: clc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001375068.3 | NP_001362078.1 | 4 - 5 | 359 | 118 | |
| NM_076856.6 | NP_509257.1 | 5 - 6 | 493 | 118 | ||
| GenBank mRNA | AF252604.1 | - | 5 - 6 | 495 | 118 | |
More Information
Set Membership: |
|

clc-2
Genomic Map