Assay Details
| Gene Symbol: clc-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076857.7 | NP_509258.1 | 1 - 2 | 182 | 107 | |
| GenBank mRNA | AF252605.1 | - | 1 - 2 | 109 | 107 | |
| Gene Symbol: clc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076856.6 | NP_509257.1 | 7 - 8 | 1171 | 107 | |
More Information
Set Membership: |
|

clc-5
Genomic Map