Assay Details
| Gene Symbol: clc-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076857.7 | NP_509258.1 | 5 - 6 | 830 | 58 | |
| GenBank mRNA | AF252605.1 | - | 5 - 6 | 757 | 58 | |
| Gene Symbol: clc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076856.6 | NP_509257.1 | 10 - 11 | 2307 | 58 | |
More Information
Set Membership: |
|

clc-5
Genomic Map