Assay Details
Gene Symbol: pcyt-2.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047761.5 | NP_001041226.2 | 7 - 8 | 698 | 65 | |
NM_001047762.5 | NP_001041227.1 | 2 - 3 | 173 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
