Assay Details
Gene Symbol: dim-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029249.6 | NP_001024420.1 | 2 - 3 | 103 | 110 | |
GenBank mRNA | AY095447.1 | - | 2 - 3 | 122 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
