Assay Details
Gene Symbol: dim-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029249.6 | NP_001024420.1 | 4 - 5 | 248 | 173 | |
GenBank mRNA | AY095447.1 | - | 4 - 5 | 267 | 173 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
