Assay Details
Gene Symbol: dim-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029249.6 | NP_001024420.1 | 6 - 7 | 949 | 81 | |
GenBank mRNA | AY095447.1 | - | 6 - 7 | 968 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
