Assay Details
Gene Symbol: ksr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076995.7 | NP_509396.1 | 6 - 7 | 809 | 122 | |
GenBank mRNA | S80647.1 | - | 6 - 7 | 830 | 122 | |
U38820.1 | - | 6 - 7 | 809 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
