Assay Details
| Gene Symbol: ksr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076995.7 | NP_509396.1 | 9 - 10 | 1433 | 95 | |
| GenBank mRNA | S80647.1 | - | 9 - 10 | 1454 | 95 | |
| U38820.1 | - | 9 - 10 | 1433 | 95 | ||
More Information
Set Membership: |
|

ksr-1
Genomic Map