Assay Details
| Gene Symbol: sprp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077098.6 | NP_509499.1 | 3 - 4 | 2109 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sprp-4| Gene Symbol: sprp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077098.6 | NP_509499.1 | 3 - 4 | 2109 | 75 | |
Set Membership: |
|