Assay Details
| Gene Symbol: nmur-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077114.4 | NP_509515.3 | 3 - 4 | 402 | 86 | |
| GenBank mRNA | HQ696948.1 | - | 3 - 4 | 402 | 86 | |
More Information
Set Membership: |
|

nmur-1
Genomic Map