Assay Details
| Gene Symbol: ajm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330906.2 | NP_001317734.1 | 20 - 21 | 4208 | 84 | |
| NM_001330908.3 | NP_001317889.1 | 14 - 15 | 3388 | 84 | ||
| NM_001330909.3 | NP_001317890.1 | 14 - 15 | 3163 | 84 | ||
| NM_001330910.3 | NP_001317891.1 | 12 - 13 | 3031 | 84 | ||
| NM_001330911.3 | NP_001317888.1 | 11 - 12 | 2917 | 84 | ||
| NM_001330912.3 | NP_001317897.1 | 10 - 11 | 2601 | 84 | ||
| NM_001330913.3 | NP_001317898.1 | 9 - 10 | 2356 | 84 | ||
| NM_001330914.3 | NP_001317899.1 | 8 - 9 | 2284 | 84 | ||
| NM_001330915.3 | NP_001317736.1 | 6 - 7 | 1987 | 84 | ||
| NM_001381045.1 | NP_001368606.1 | 20 - 21 | 4171 | 84 | ||
| NM_001381048.1 | NP_001367381.1 | 20 - 21 | 4208 | 84 | ||
More Information
Set Membership: |
|

ajm-1
Genomic Map