Assay Details
| Gene Symbol: vav-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047757.3 | NP_001041222.1 | 2 - 3 | 206 | 150 | |
| NM_001373412.3 | NP_001359918.1 | 3 - 4 | 362 | 150 | ||
| NM_001373413.3 | NP_001360680.1 | 3 - 4 | 349 | 150 | ||
| GenBank mRNA | DQ136172.1 | - | 3 - 4 | 300 | 150 | |
More Information
Set Membership: |
|

vav-1
Genomic Map