Assay Details
| Gene Symbol: vav-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047757.3 | NP_001041222.1 | 6 - 7 | 917 | 93 | |
| NM_001373412.3 | NP_001359918.1 | 7 - 8 | 1073 | 93 | ||
| NM_001373413.3 | NP_001360680.1 | 7 - 8 | 1060 | 93 | ||
| GenBank mRNA | DQ136172.1 | - | 7 - 8 | 1011 | 93 | |
More Information
Set Membership: |
|

vav-1
Genomic Map