Assay Details
| Gene Symbol: him-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029411.3 | NP_001024582.1 | 59 - 60 | 15158 | 64 | |
| NM_001129660.3 | NP_001123132.1 | 1 - 2 | 139 | 64 | ||
| NM_001373450.2 | NP_001360633.1 | 58 - 59 | 14803 | 64 | ||
| NM_001373451.2 | NP_001360634.1 | 58 - 59 | 14734 | 64 | ||
| NM_077234.7 | NP_509635.2 | 59 - 60 | 15028 | 64 | ||
| GenBank mRNA | AF074901.1 | - | 59 - 60 | 15097 | 64 | |
More Information
Set Membership: |
|

him-4
Genomic Map