Assay Details
Gene Symbol: mkk-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077281.7 | NP_509682.1 | 4 - 5 | 536 | 158 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
