Assay Details
Gene Symbol: flp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077293.8 | NP_509694.1 | 1 - 2 | 113 | 82 | |
GenBank mRNA | AF042389.1 | - | 1 - 2 | 67 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
