Assay Details
Gene Symbol: flp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077293.8 | NP_509694.1 | 2 - 3 | 305 | 83 | |
GenBank mRNA | AF042389.1 | - | 2 - 3 | 259 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
