Assay Details
| Gene Symbol: acr-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001375112.2 | NP_001361818.1 | 9 | 1104 | 143 | |
| GenBank mRNA | MF767409.1 | - | 9 - 10 | 1025 | 143 | |
More Information
Set Membership: |
|

acr-8
Genomic Map