Assay Details
Gene Symbol: acr-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375112.2 | NP_001361818.1 | 10 | 1228 | 69 | |
GenBank mRNA | MF767409.1 | - | 10 - 11 | 1149 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
