Assay Details
| Gene Symbol: mboa-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077359.7 | NP_509760.2 | 3 - 4 | 364 | 75 | |
| GenBank mRNA | EU016382.1 | - | 3 - 4 | 356 | 75 | |
More Information
Set Membership: |
|

mboa-7
Genomic Map