Assay Details
| Gene Symbol: mboa-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077359.7 | NP_509760.2 | 4 - 5 | 502 | 153 | |
| GenBank mRNA | EU016382.1 | - | 4 - 5 | 494 | 153 | |
More Information
Set Membership: |
|

mboa-7
Genomic Map