Assay Details
Gene Symbol: mbr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077393.7 | NP_509794.2 | 2 - 3 | 188 | 64 | |
GenBank mRNA | AB236333.1 | - | 1 - 2 | 92 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
