Assay Details
Gene Symbol: mbr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077393.7 | NP_509794.2 | 4 - 5 | 430 | 85 | |
GenBank mRNA | AB236333.1 | - | 3 - 4 | 334 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
