Assay Details
| Gene Symbol: plc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029446.3 | NP_001024617.1 | 12 - 13 | 2292 | 129 | |
| NM_001029448.6 | NP_001024619.1 | 12 - 13 | 2254 | 129 | ||
| NM_001129669.4 | NP_001123141.2 | 45 - 46 | 7132 | 129 | ||
| NM_001136454.3 | NP_001129926.1 | 12 - 13 | 2284 | 129 | ||
| NM_001136455.4 | NP_001129927.1 | 12 - 13 | 2476 | 129 | ||
| NM_001373489.2 | NP_001360637.1 | 45 - 46 | 7111 | 129 | ||
| NM_001373491.2 | NP_001360639.1 | 45 - 46 | 7141 | 129 | ||
| NM_001373492.2 | NP_001360635.1 | 45 - 46 | 7162 | 129 | ||
| NM_001373493.2 | NP_001360636.1 | 46 - 47 | 7282 | 129 | ||
| NM_001373494.2 | NP_001359720.1 | 46 - 47 | 7303 | 129 | ||
| NM_001373495.2 | NP_001360638.1 | 46 - 47 | 7312 | 129 | ||
| NM_077404.6 | NP_509805.3 | 12 - 13 | 2254 | 129 | ||
| GenBank mRNA | AF044576.1 | - | 12 - 13 | 2314 | 129 | |
| AM992255.1 | - | 12 - 13 | 2284 | 129 | ||
More Information
Set Membership: |
|

plc-1
Genomic Map