Assay Details
| Gene Symbol: plc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029446.3 | NP_001024617.1 | 19 - 20 | 3331 | 96 | |
| NM_001029448.6 | NP_001024619.1 | 19 - 20 | 3293 | 96 | ||
| NM_001129669.4 | NP_001123141.2 | 52 - 53 | 8171 | 96 | ||
| NM_001136454.3 | NP_001129926.1 | 19 - 20 | 3323 | 96 | ||
| NM_001136455.4 | NP_001129927.1 | 19 - 20 | 3515 | 96 | ||
| NM_001373489.2 | NP_001360637.1 | 52 - 53 | 8150 | 96 | ||
| NM_001373491.2 | NP_001360639.1 | 52 - 53 | 8180 | 96 | ||
| NM_001373492.2 | NP_001360635.1 | 52 - 53 | 8201 | 96 | ||
| NM_001373493.2 | NP_001360636.1 | 53 - 54 | 8321 | 96 | ||
| NM_001373494.2 | NP_001359720.1 | 53 - 54 | 8342 | 96 | ||
| NM_001373495.2 | NP_001360638.1 | 53 - 54 | 8351 | 96 | ||
| NM_077404.6 | NP_509805.3 | 19 - 20 | 3293 | 96 | ||
| GenBank mRNA | AF044576.1 | - | 19 - 20 | 3353 | 96 | |
| AM992255.1 | - | 19 - 20 | 3323 | 96 | ||
More Information
Set Membership: |
|

plc-1
Genomic Map