Assay Details
| Gene Symbol: plc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029446.3 | NP_001024617.1 | 4 - 5 | 891 | 81 | |
| NM_001029448.6 | NP_001024619.1 | 4 - 5 | 853 | 81 | ||
| NM_001129669.4 | NP_001123141.2 | 37 - 38 | 5731 | 81 | ||
| NM_001136454.3 | NP_001129926.1 | 4 - 5 | 853 | 81 | ||
| NM_001136455.4 | NP_001129927.1 | 4 - 5 | 1075 | 81 | ||
| NM_001373489.2 | NP_001360637.1 | 37 - 38 | 5710 | 81 | ||
| NM_001373491.2 | NP_001360639.1 | 37 - 38 | 5740 | 81 | ||
| NM_001373492.2 | NP_001360635.1 | 37 - 38 | 5761 | 81 | ||
| NM_001373493.2 | NP_001360636.1 | 38 - 39 | 5881 | 81 | ||
| NM_001373494.2 | NP_001359720.1 | 38 - 39 | 5902 | 81 | ||
| NM_001373495.2 | NP_001360638.1 | 38 - 39 | 5911 | 81 | ||
| NM_077404.6 | NP_509805.3 | 4 - 5 | 853 | 81 | ||
| GenBank mRNA | AF044576.1 | - | 4 - 5 | 913 | 81 | |
| AM992255.1 | - | 4 - 5 | 853 | 81 | ||
More Information
Set Membership: |
|

plc-1
Genomic Map