Assay Details
| Gene Symbol: ver-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077435.6 | NP_509836.1 | 6 - 7 | 1962 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ver-3| Gene Symbol: ver-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077435.6 | NP_509836.1 | 6 - 7 | 1962 | 70 | |
Set Membership: |
|