Assay Details
Gene Symbol: ldb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381099.1 | NP_001366942.1 | 5 - 6 | 776 | 88 | |
NM_001381100.2 | NP_001366860.1 | 6 - 7 | 1006 | 88 | ||
NM_077444.6 | NP_509845.1 | 5 - 6 | 733 | 88 | ||
GenBank mRNA | U80220.1 | - | 4 - 5 | 665 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
