Assay Details
| Gene Symbol: gcy-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077496.5 | NP_509897.3 | 4 - 5 | 735 | 99 | |
| GenBank mRNA | HQ636455.1 | - | 3 - 4 | 344 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gcy-9| Gene Symbol: gcy-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077496.5 | NP_509897.3 | 4 - 5 | 735 | 99 | |
| GenBank mRNA | HQ636455.1 | - | 3 - 4 | 344 | 99 | |
Set Membership: |
|