Assay Details
| Gene Symbol: slo-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029356.4 | NP_001024527.1 | 15 - 16 | 3271 | 123 | |
| NM_001029357.5 | NP_001024528.1 | 15 - 16 | 3271 | 123 | ||
| NM_001029358.5 | NP_001024529.1 | 15 - 16 | 2952 | 123 | ||
| NM_001136452.4 | NP_001129924.1 | 15 - 16 | 2955 | 123 | ||
| NM_001270215.3 | NP_001257144.1 | 17 - 18 | 3063 | 123 | ||
| NM_001270216.3 | NP_001257145.1 | 16 - 17 | 3018 | 123 | ||
| NM_001270217.3 | NP_001257146.1 | 16 - 17 | 3072 | 123 | ||
| NM_001270218.3 | NP_001257147.1 | 16 - 17 | 3033 | 123 | ||
| GenBank mRNA | AF173828.1 | - | 15 - 16 | 3163 | 123 | |
| AF232770.1 | - | 15 - 16 | 2964 | 123 | ||
More Information
Set Membership: |
|

slo-2
Genomic Map