Assay Details
| Gene Symbol: slo-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029356.4 | NP_001024527.1 | 16 - 17 | 3397 | 135 | |
| NM_001136452.4 | NP_001129924.1 | 16 - 17 | 3081 | 135 | ||
| GenBank mRNA | AF173828.1 | - | 16 - 17 | 3289 | 135 | |
More Information
Set Membership: |
|

slo-2
Genomic Map