Assay Details
| Gene Symbol: slo-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029356.4 | NP_001024527.1 | 4 - 5 | 752 | 117 | |
| NM_001029357.5 | NP_001024528.1 | 4 - 5 | 752 | 117 | ||
| NM_001029358.5 | NP_001024529.1 | 4 - 5 | 433 | 117 | ||
| NM_001136452.4 | NP_001129924.1 | 4 - 5 | 436 | 117 | ||
| NM_001270215.3 | NP_001257144.1 | 6 - 7 | 544 | 117 | ||
| NM_001270216.3 | NP_001257145.1 | 5 - 6 | 499 | 117 | ||
| NM_001270217.3 | NP_001257146.1 | 5 - 6 | 553 | 117 | ||
| NM_001270218.3 | NP_001257147.1 | 5 - 6 | 514 | 117 | ||
| GenBank mRNA | AF173828.1 | - | 4 - 5 | 644 | 117 | |
| AF232770.1 | - | 4 - 5 | 445 | 117 | ||
More Information
Set Membership: |
|

slo-2
Genomic Map