Assay Details
| Gene Symbol: slo-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029356.4 | NP_001024527.1 | 5 - 6 | 822 | 119 | |
| NM_001029357.5 | NP_001024528.1 | 5 - 6 | 822 | 119 | ||
| NM_001029358.5 | NP_001024529.1 | 5 - 6 | 503 | 119 | ||
| NM_001136452.4 | NP_001129924.1 | 5 - 6 | 506 | 119 | ||
| NM_001270215.3 | NP_001257144.1 | 7 - 8 | 614 | 119 | ||
| NM_001270216.3 | NP_001257145.1 | 6 - 7 | 569 | 119 | ||
| NM_001270217.3 | NP_001257146.1 | 6 - 7 | 623 | 119 | ||
| NM_001270218.3 | NP_001257147.1 | 6 - 7 | 584 | 119 | ||
| GenBank mRNA | AF173828.1 | - | 5 - 6 | 714 | 119 | |
| AF232770.1 | - | 5 - 6 | 515 | 119 | ||
More Information
Set Membership: |
|

slo-2
Genomic Map