Assay Details
| Gene Symbol: pek-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077511.8 | NP_509912.1 | 7 - 8 | 1698 | 96 | |
| GenBank mRNA | AF193341.1 | - | 7 - 8 | 1622 | 96 | |
| AF435953.1 | - | 7 - 8 | 1622 | 96 | ||
More Information
Set Membership: |
|

pek-1
Genomic Map