Assay Details
| Gene Symbol: tag-241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270245.2 | NP_001257174.1 | 4 - 5 | 557 | 72 | |
| NM_001368527.5 | NP_001355535.1 | 4 - 5 | 421 | 72 | ||
| NM_001368528.3 | NP_001355536.1 | 4 - 5 | 413 | 72 | ||
| NM_001368529.3 | NP_001355537.1 | 4 - 5 | 413 | 72 | ||
| NM_001381110.1 | NP_001367232.1 | 3 - 4 | 374 | 72 | ||
| NM_001383651.1 | 4 - 5 | 429 | 72 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383651.1 transcripts.
Set Membership: |
|

tag-241
Genomic Map