Assay Details
| Gene Symbol: tag-241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270245.2 | NP_001257174.1 | 7 - 8 | 1272 | 63 | |
| NM_001368527.5 | NP_001355535.1 | 7 - 8 | 1136 | 63 | ||
| NM_001368528.3 | NP_001355536.1 | 7 - 8 | 1128 | 63 | ||
| NM_001368529.3 | NP_001355537.1 | 7 - 8 | 1128 | 63 | ||
| NM_001375149.2 | NP_001361908.1 | 7 - 8 | 1145 | 63 | ||
| NM_001381110.1 | NP_001367232.1 | 6 - 7 | 1089 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001375149.2 transcripts.
Set Membership: |
|

tag-241
Genomic Map