Assay Details
| Gene Symbol: lin-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029416.4 | NP_001024587.1 | 10 - 11 | 1129 | 62 | |
| GenBank mRNA | X92564.1 | - | 1113 | 62 | ||
More Information
Set Membership: |
|

lin-2
Genomic Map